Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439275.1 | XP_015294761.1 | 5 - 6 | 2382 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCRTR1| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439275.1 | XP_015294761.1 | 5 - 6 | 2382 | 69 | |
Set Membership: |
|