Assay Details
| Gene Symbol: TNFRSF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544888.2 | XP_005544945.1 | 5 - 6 | 1245 | 147 | |
| XM_005544890.2 | XP_005544947.1 | 5 - 6 | 1195 | 147 | ||
| XM_005544891.2 | XP_005544948.1 | 5 - 6 | 1182 | 147 | ||
| GenBank mRNA | EU310931.1 | - | 4 - 5 | 428 | 147 | |
More Information
Set Membership: |
|

TNFRSF9
Genomic Map