Assay Details
| Gene Symbol: ACADL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285823.1 | NP_001272752.1 | 8 - 9 | 1101 | 129 | |
| XM_005574160.2 | XP_005574217.1 | 8 - 9 | 1647 | 129 | ||
| GenBank mRNA | AB125147.1 | - | 8 - 9 | 1101 | 129 | |
| AB174488.1 | - | 8 - 9 | 1101 | 129 | ||
More Information
Set Membership: |
|

ACADL
Genomic Map