Assay Details
| Gene Symbol: AREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555060.2 | XP_005555117.1 | 3 - 4 | 764 | 77 | |
| XM_005555061.2 | XP_005555118.1 | 3 - 4 | 610 | 77 | ||
| XM_005555062.2 | XP_005555119.1 | 3 - 4 | 735 | 77 | ||
More Information
Set Membership: |
|

AREG
Genomic Map