Assay Details
| Gene Symbol: RUNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548734.2 | XP_005548791.1 | 8 - 9 | 1204 | 68 | |
| XM_005548743.2 | XP_005548800.1 | 7 - 8 | 1165 | 68 | ||
| XM_005548745.2 | XP_005548802.1 | 7 - 8 | 1012 | 68 | ||
| XM_005548746.2 | XP_005548803.1 | 6 - 7 | 973 | 68 | ||
More Information
Set Membership: |
|

RUNX1
Genomic Map