Assay Details
| Gene Symbol: LOC102123466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545062.2 | XP_005545119.1 | 11 - 12 | 2480 | 140 | |
| XM_005545063.2 | XP_005545120.1 | 11 - 12 | 2450 | 140 | ||
| XM_005545064.2 | XP_005545121.1 | 12 - 13 | 1489 | 140 | ||
| XM_005545065.2 | XP_005545122.1 | 12 - 13 | 2497 | 140 | ||
| XM_015443840.1 | XP_015299326.1 | 11 - 12 | 1552 | 140 | ||
| XM_015443842.1 | XP_015299328.1 | 11 - 12 | 1524 | 140 | ||
| XM_015443844.1 | XP_015299330.1 | 11 - 12 | 1431 | 140 | ||
| XM_015443850.1 | XP_015299336.1 | 10 - 11 | 1305 | 140 | ||
| XM_015443852.1 | XP_015299338.1 | 10 - 11 | 2313 | 140 | ||
More Information
Set Membership: |
|

LOC102123466
Genomic Map