Assay Details
| Gene Symbol: CLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562916.2 | XP_005562973.1 | 6 - 7 | 1332 | 144 | |
| XM_005562917.2 | XP_005562974.1 | 6 - 7 | 1250 | 144 | ||
| XM_015454612.1 | XP_015310098.1 | 6 - 7 | 1573 | 144 | ||
| GenBank mRNA | AB171269.1 | - | 5 - 6 | 671 | 144 | |
| AB171907.1 | - | 2 - 3 | 532 | 144 | ||
| AY650328.1 | - | 5 - 6 | 870 | 144 | ||
More Information
Set Membership: |
|

CLU
Genomic Map