Assay Details
| Gene Symbol: LOC102144252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567301.2 | XP_005567358.2 | 2 - 3 | 389 | 90 | |
| XM_005567302.2 | XP_005567359.1 | 2 - 3 | 208 | 90 | ||
| XM_005567303.1 | XP_005567360.1 | 2 - 3 | 208 | 90 | ||
| XM_005567304.2 | XP_005567361.1 | 2 - 3 | 208 | 90 | ||
| XM_015457211.1 | XP_015312697.1 | 2 - 3 | 170 | 90 | ||
More Information
Set Membership: |
|

LOC102144252
Genomic Map