Assay Details
| Gene Symbol: FKBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569814.2 | XP_005569871.1 | 4 - 5 | 708 | 122 | |
| GenBank mRNA | AB169439.1 | - | 1 - 2 | 283 | 122 | |
More Information
Set Membership: |
|

FKBP4
Genomic Map