Assay Details
| Gene Symbol: KCNN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589466.2 | XP_005589523.1 | 7 - 8 | 3540 | 66 | |
| XM_005589467.2 | XP_005589524.1 | 6 - 7 | 3445 | 66 | ||
| XM_005589470.2 | XP_005589527.1 | 7 - 8 | 1664 | 66 | ||
| XM_015441885.1 | XP_015297371.1 | 7 - 8 | 3508 | 66 | ||
| XM_015441886.1 | XP_015297372.1 | 7 - 8 | 1519 | 66 | ||
| XM_015441887.1 | XP_015297373.1 | 7 - 8 | 1660 | 66 | ||
More Information
Set Membership: |
|

KCNN4
Genomic Map