Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553305.2 | XP_005553362.2 | 7 - 8 | 2041 | 72 | |
| XM_005553307.2 | XP_005553364.1 | 7 - 8 | 1828 | 72 | ||
| XM_015449554.1 | XP_015305040.1 | 7 - 8 | 1937 | 72 | ||
More Information
Set Membership: |
|

KIFC1
Genomic Map