Assay Details
| Gene Symbol: LOC102120872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570926.1 | XP_005570983.1 | 8 - 9 | 1455 | 122 | |
| XM_015430958.1 | XP_015286444.1 | 6 - 7 | 864 | 122 | ||
More Information
Set Membership: |
|

LOC102120872
Genomic Map