Assay Details
| Gene Symbol: MMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579466.2 | XP_005579523.1 | 8 - 9 | 1313 | 119 | |
| XM_005579467.2 | XP_005579524.1 | 8 - 9 | 3099 | 119 | ||
| XM_015435640.1 | XP_015291126.1 | 9 - 10 | 1385 | 119 | ||
More Information
Set Membership: |
|

MMP8
Genomic Map