Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557097.2 | XP_005557154.1 | 3 - 4 | 1162 | 145 | |
| XM_005557098.2 | XP_005557155.1 | 3 - 4 | 939 | 145 | ||
| XM_015451590.1 | XP_015307076.1 | 3 - 4 | 876 | 145 | ||
| XM_015451591.1 | XP_015307077.1 | 2 - 3 | 810 | 145 | ||
More Information
Set Membership: |
|

OCLN
Genomic Map