Assay Details
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570879.2 | XP_005570936.1 | 10 - 11 | 1767 | 83 | |
XM_015430929.1 | XP_015286415.1 | 10 - 11 | 1922 | 83 | ||
XM_015430930.1 | XP_015286416.1 | 11 - 12 | 2039 | 83 | ||
XM_015430931.1 | XP_015286417.1 | 10 - 11 | 1919 | 83 | ||
XM_015430932.1 | XP_015286418.1 | 9 - 10 | 1793 | 83 | ||
XM_015430933.1 | XP_015286419.1 | 9 - 10 | 1739 | 83 | ||
XM_015430934.1 | XP_015286420.1 | 10 - 11 | 1862 | 83 | ||
XM_015430935.1 | XP_015286421.1 | 8 - 9 | 1374 | 83 | ||
XM_015430936.1 | XP_015286422.1 | 6 - 7 | 1042 | 83 | ||
XM_015430937.1 | XP_015286423.1 | 9 - 10 | 1637 | 83 | ||
GenBank mRNA | AB171729.1 | - | 4 - 5 | 662 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
