Assay Details
| Gene Symbol: REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575801.2 | XP_005575858.1 | 7 - 8 | 1190 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

REL| Gene Symbol: REL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575801.2 | XP_005575858.1 | 7 - 8 | 1190 | 124 | |
Set Membership: |
|