Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557722.2 | XP_005557779.1 | 2 - 3 | 774 | 81 | |
| XM_015451865.1 | XP_015307351.1 | 3 - 4 | 846 | 81 | ||
More Information
Set Membership: |
|

SLC22A5
Genomic Map