Assay Details
| Gene Symbol: SPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577948.2 | XP_005578005.1 | 1 - 2 | 259 | 63 | |
| XM_005577950.2 | XP_005578007.1 | 1 - 2 | 268 | 63 | ||
More Information
Set Membership: |
|

SPI1
Genomic Map