Assay Details
| Gene Symbol: STAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545470.2 | XP_005545527.1 | 3 - 4 | 790 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STAC| Gene Symbol: STAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545470.2 | XP_005545527.1 | 3 - 4 | 790 | 82 | |
Set Membership: |
|