Assay Details
| Gene Symbol: TGFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561843.2 | XP_005561900.1 | 7 - 8 | 2456 | 133 | |
| XM_015454019.1 | XP_015309505.1 | 6 - 7 | 2709 | 133 | ||
More Information
Set Membership: |
|

TGFB3
Genomic Map