Assay Details
| Gene Symbol: GPR137B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539723.2 | XP_005539780.1 | 2 - 3 | 475 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR137B| Gene Symbol: GPR137B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539723.2 | XP_005539780.1 | 2 - 3 | 475 | 101 | |
Set Membership: |
|