Assay Details
| Gene Symbol: UBE2G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548528.2 | XP_005548585.1 | 5 - 6 | 489 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2G2| Gene Symbol: UBE2G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548528.2 | XP_005548585.1 | 5 - 6 | 489 | 125 | |
Set Membership: |
|