Assay Details
| Gene Symbol: SUPT3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552831.2 | XP_005552888.1 | 2 - 3 | 1290 | 111 | |
| XM_005552832.2 | XP_005552889.1 | 3 - 4 | 527 | 111 | ||
| XM_005552834.2 | XP_005552891.1 | 2 - 3 | 249 | 111 | ||
| XM_005552836.2 | XP_005552893.1 | 2 - 3 | 2578 | 111 | ||
| XM_005552839.2 | XP_005552896.1 | 2 - 3 | 380 | 111 | ||
| XM_015449351.1 | XP_015304837.1 | 3 - 4 | 343 | 111 | ||
More Information
Set Membership: |
|

SUPT3H
Genomic Map