Assay Details
| Gene Symbol: LOC102119459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570431.2 | XP_005570488.1 | 13 - 14 | 1458 | 92 | |
| XM_005570433.2 | XP_005570490.1 | 14 - 15 | 1468 | 92 | ||
| XM_005570439.1 | XP_005570496.1 | 12 - 13 | 1370 | 92 | ||
| XM_015430553.1 | XP_015286039.1 | 14 - 15 | 1545 | 92 | ||
| XM_015430554.1 | XP_015286040.1 | 14 - 15 | 1512 | 92 | ||
| XM_015430555.1 | XP_015286041.1 | 15 - 16 | 1608 | 92 | ||
| XM_015430557.1 | XP_015286043.1 | 13 - 14 | 1512 | 92 | ||
| XM_015430558.1 | XP_015286044.1 | 14 - 15 | 1545 | 92 | ||
| XM_015430559.1 | XP_015286045.1 | 13 - 14 | 1512 | 92 | ||
| XM_015430561.1 | XP_015286047.1 | 13 - 14 | 1452 | 92 | ||
| XM_015430563.1 | XP_015286049.1 | 11 - 12 | 1141 | 92 | ||
| XM_015430564.1 | XP_015286050.1 | 11 - 12 | 1133 | 92 | ||
| XM_015430565.1 | XP_015286051.1 | 12 - 13 | 1419 | 92 | ||

LOC102119459
Genomic Map