Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539920.2 | XP_005539977.1 | 4 - 5 | 1069 | 94 | |
| XM_005539921.2 | XP_005539978.1 | 3 - 4 | 728 | 94 | ||
More Information
Set Membership: |
|

BLZF1
Genomic Map