Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574079.2 | XP_005574136.1 | 1 - 2 | 717 | 142 | |
| XM_005574080.2 | XP_005574137.1 | 2 - 3 | 508 | 142 | ||
| XM_005574081.2 | XP_005574138.1 | 1 - 2 | 717 | 142 | ||
| XM_005574083.2 | XP_005574140.1 | 1 - 2 | 717 | 142 | ||
More Information
Set Membership: |
|

KLF7
Genomic Map