Assay Details
| Gene Symbol: B4GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548073.2 | XP_005548130.1 | 6 - 7 | 1276 | 127 | |
| XM_005548074.2 | XP_005548131.1 | 5 - 6 | 1042 | 127 | ||
| XM_015446392.1 | XP_015301878.1 | 6 - 7 | 1430 | 127 | ||
| XM_015446393.1 | XP_015301879.1 | 4 - 5 | 564 | 127 | ||
More Information
Set Membership: |
|

B4GALT4
Genomic Map