Assay Details
| Gene Symbol: TNFRSF10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454407.1 | XP_015309893.1 | 6 - 7 | 619 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF10A| Gene Symbol: TNFRSF10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454407.1 | XP_015309893.1 | 6 - 7 | 619 | 113 | |
Set Membership: |
|