Assay Details
| Gene Symbol: PROZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586293.2 | XP_005586350.1 | 5 - 6 | 712 | 107 | |
| XM_005586295.2 | XP_005586352.1 | 5 - 6 | 662 | 107 | ||
More Information
Set Membership: |
|

PROZ
Genomic Map