Assay Details
| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583077.2 | XP_005583134.1 | 4 - 5 | 579 | 98 | |
| XM_005583078.2 | XP_005583135.1 | 4 - 5 | 508 | 98 | ||
| XM_015437742.1 | XP_015293228.1 | 4 - 5 | 740 | 98 | ||
More Information
Set Membership: |
|

SHMT1
Genomic Map