Assay Details
| Gene Symbol: FADS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577619.2 | XP_005577676.1 | 1 - 2 | 834 | 82 | |
| GenBank mRNA | AB168976.1 | - | 2 - 3 | 316 | 82 | |
More Information
Set Membership: |
|

FADS2
Genomic Map