Assay Details
| Gene Symbol: BNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559688.2 | XP_005559745.1 | 5 - 6 | 1433 | 141 | |
| XM_005559689.2 | XP_005559746.1 | 5 - 6 | 1433 | 141 | ||
More Information
Set Membership: |
|

BNIP2
Genomic Map