Assay Details
| Gene Symbol: KCTD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572183.2 | XP_005572240.1 | 5 - 6 | 1070 | 104 | |
| XM_005572186.2 | XP_005572243.1 | 5 - 6 | 811 | 104 | ||
| XM_005572187.2 | XP_005572244.2 | 5 - 6 | 640 | 104 | ||
| XM_005572189.2 | XP_005572246.1 | 5 - 6 | 814 | 104 | ||
| XM_015431507.1 | XP_015286993.1 | 5 - 6 | 632 | 104 | ||
More Information
Set Membership: |
|

KCTD10
Genomic Map