Assay Details
| Gene Symbol: RASSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565101.2 | XP_005565158.1 | 2 - 3 | 211 | 81 | |
| XM_005565102.2 | XP_005565159.1 | 2 - 3 | 193 | 81 | ||
| XM_005565103.2 | XP_005565160.1 | 2 - 3 | 164 | 81 | ||
More Information
Set Membership: |
|

RASSF4
Genomic Map