Assay Details
| Gene Symbol: C19H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588332.2 | XP_005588389.1 | 2 - 3 | 906 | 155 | |
| XM_015440611.1 | XP_015296097.1 | 2 - 3 | 906 | 155 | ||
| XM_015440612.1 | XP_015296098.1 | 2 - 3 | 831 | 155 | ||
| XM_015440613.1 | XP_015296099.1 | 2 - 3 | 835 | 155 | ||
| XM_015440614.1 | XP_015296100.1 | 2 - 3 | 906 | 155 | ||
| XM_015440615.1 | XP_015296101.1 | 2 - 3 | 906 | 155 | ||
| XM_015440616.1 | XP_015296102.1 | 2 - 3 | 906 | 155 | ||
| GenBank mRNA | AB168765.1 | - | 517 | 155 | ||
More Information
Set Membership: |
|

C19H19orf44
Genomic Map