Assay Details
| Gene Symbol: SLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568931.2 | XP_005568988.1 | 5 - 6 | 1500 | 122 | |
| XM_005568932.2 | XP_005568989.1 | 6 - 7 | 1588 | 122 | ||
| XM_015458028.1 | XP_015313514.1 | 5 - 6 | 1501 | 122 | ||
More Information
Set Membership: |
|

SLA2
Genomic Map