Assay Details
| Gene Symbol: CCDC88B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577414.2 | XP_005577471.2 | 7 - 8 | 1468 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC88B| Gene Symbol: CCDC88B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577414.2 | XP_005577471.2 | 7 - 8 | 1468 | 96 | |
Set Membership: |
|