Assay Details
| Gene Symbol: EIF1AD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283992.1 | NP_001270921.1 | 2 - 3 | 576 | 110 | |
| XM_005577197.2 | XP_005577254.1 | 2 - 3 | 670 | 110 | ||
| GenBank mRNA | AB179414.1 | - | 2 - 3 | 576 | 110 | |
More Information
Set Membership: |
|

EIF1AD
Genomic Map