Assay Details
| Gene Symbol: LOC102124387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544716.2 | XP_005544773.1 | 5 - 6 | 968 | 68 | |
| XM_005544718.2 | XP_005544775.1 | 5 - 6 | 969 | 68 | ||
| XM_005544719.2 | XP_005544776.1 | 5 - 6 | 968 | 68 | ||
| XM_015442430.1 | XP_015297916.1 | 1 - 2 | 31 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015442430.1 transcripts.
Set Membership: |
|

LOC102124387
Genomic Map