Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287689.1 | NP_001274618.1 | 4 - 5 | 384 | 84 | |
| XM_005558362.2 | XP_005558419.1 | 4 - 5 | 675 | 84 | ||
| XM_005558363.2 | XP_005558420.1 | 4 - 5 | 357 | 84 | ||
| GenBank mRNA | AB052147.1 | - | 4 - 5 | 384 | 84 | |
| AB056418.1 | - | 4 - 5 | 384 | 84 | ||
More Information
Set Membership: |
|

FAXDC2
Genomic Map