Assay Details
| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564164.2 | XP_005564221.1 | 3 - 4 | 1175 | 117 | |
| XM_005564165.2 | XP_005564222.1 | 3 - 4 | 1161 | 117 | ||
| XM_015455262.1 | XP_015310748.1 | 3 - 4 | 1135 | 117 | ||
| XM_015455264.1 | XP_015310750.1 | 2 - 3 | 2243 | 117 | ||
| XM_015455265.1 | XP_015310751.1 | 3 - 4 | 1175 | 117 | ||
More Information
Set Membership: |
|

PTP4A3
Genomic Map