Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587385.2 | XP_005587442.1 | 8 - 9 | 1563 | 124 | |
| XM_005587386.2 | XP_005587443.1 | 9 - 10 | 1579 | 124 | ||
| XM_005587387.1 | XP_005587444.1 | 9 - 10 | 1562 | 124 | ||
| XM_015440321.1 | XP_015295807.1 | 8 - 9 | 1449 | 124 | ||
| XM_015440323.1 | XP_015295809.1 | 9 - 10 | 1555 | 124 | ||
| XM_015440324.1 | XP_015295810.1 | 5 - 6 | 1471 | 124 | ||
| XM_015440325.1 | XP_015295811.1 | 2 - 3 | 181 | 124 | ||
More Information
Set Membership: |
|

PWWP3A
Genomic Map