Assay Details
| Gene Symbol: TOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569053.2 | XP_005569110.2 | 5 - 6 | 1471 | 113 | |
| XM_005569054.2 | XP_005569111.1 | 5 - 6 | 810 | 113 | ||
| XM_015429861.1 | XP_015285347.1 | 5 - 6 | 1471 | 113 | ||
More Information
Set Membership: |
|

TOX2
Genomic Map