Assay Details
| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443371.1 | XP_015298857.1 | 3 - 4 | 1070 | 152 | |
| XM_015443372.1 | XP_015298858.1 | 3 - 4 | 1086 | 155 | ||
More Information
Set Membership: |
|

PPP1R3F
Genomic Map