Assay Details
| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559177.2 | XP_005559234.1 | 4 - 5 | 595 | 135 | |
| XM_005559178.2 | XP_005559235.1 | 4 - 5 | 595 | 135 | ||
More Information
Set Membership: |
|

KNSTRN
Genomic Map