Assay Details
| Gene Symbol: TRIM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558837.2 | XP_005558894.1 | 2 - 3 | 2392 | 75 | |
| XM_005558838.2 | XP_005558895.1 | 2 - 3 | 2097 | 75 | ||
| XM_005558839.2 | XP_005558896.1 | 2 - 3 | 2096 | 75 | ||
| XM_005558840.2 | XP_005558897.1 | 2 - 3 | 2092 | 75 | ||
| XM_005558841.2 | XP_005558898.1 | 2 - 3 | 2093 | 75 | ||
| XM_005558842.2 | XP_005558899.1 | 2 - 3 | 2093 | 75 | ||
More Information
Set Membership: |
|

TRIM41
Genomic Map