Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591652.2 | XP_005591709.1 | 10 - 11 | 1077 | 112 | |
| XM_005591654.2 | XP_005591711.1 | 8 - 9 | 1107 | 112 | ||
| XM_015442486.1 | XP_015297972.1 | 9 - 10 | 1209 | 112 | ||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map