Assay Details
| Gene Symbol: CTCFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569424.1 | XP_005569481.1 | 6 - 7 | 1850 | 67 | |
| XM_005569425.2 | XP_005569482.1 | 6 - 7 | 1385 | 67 | ||
| XM_005569427.2 | XP_005569484.1 | 6 - 7 | 1385 | 67 | ||
More Information
Set Membership: |
|

CTCFL
Genomic Map