Assay Details
| Gene Symbol: C10H20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568829.2 | XP_005568886.1 | 3 - 4 | 630 | 75 | |
| XM_015457996.1 | XP_015313482.1 | 2 - 3 | 638 | 75 | ||
More Information
Set Membership: |
|

C10H20orf173
Genomic Map