Assay Details
Gene Symbol: IRAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578616.2 | XP_005578673.1 | 12 - 13 | 2040 | 67 | |
XM_005578619.2 | XP_005578676.1 | 12 - 13 | 1621 | 67 | ||
XM_005578621.2 | XP_005578678.1 | 11 - 12 | 1445 | 67 | ||
XM_005578622.2 | XP_005578679.1 | 11 - 12 | 1447 | 67 | ||
XM_015435119.1 | XP_015290605.1 | 11 - 12 | 1593 | 67 | ||
XM_015435120.1 | XP_015290606.1 | 11 - 12 | 11319 | 67 | ||
XM_015435121.1 | XP_015290607.1 | 11 - 12 | 11316 | 67 | ||
XM_015435122.1 | XP_015290608.1 | 11 - 12 | 11313 | 67 | ||
XM_015435123.1 | XP_015290609.1 | 11 - 12 | 1596 | 67 | ||
XM_015435124.1 | XP_015290610.1 | 11 - 12 | 1503 | 67 | ||
XM_015435125.1 | XP_015290611.1 | 11 - 12 | 1885 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
